Order Entry
Czech Republic
ContactUsLinkComponent
 
Water, PCR grade
Catalog # 733-2573
Supplier:  VWR Chemicals
CAS Number:  
undefined
Water, PCR grade
Catalog # 733-2573
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic tube
  • Pk:
    1 KIT
  • Seq:
    2345
  • Pk Info:
    6×5 ml

Specifications

Specification Test Results

  • DNA nicking activity:
    Passed
  • Endonuclease activity:
    Passed
  • Exonuclease activity:
    Passed
  • Absence of Human DNA:
    Passed
  • Volume check, 5 ml/tube:
    Passed

Specification Test Results

About this item

 :
 :18.02 g/mol
Boiling Pt:100 °C
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Storage Temperature:Freezer
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332