ContactUsLinkComponent
Rechargeable safety torch, Atex zone 1
Rechargeable safety torch, Atex zone 1
  SIMEDI0070
undefined
Rechargeable safety torch, Atex zone 1
  SIMEDI0070
  • Description:
    LED rechargeable flashlight, Atex zone 1 with scratch proof PC lens and safety switch, NiMH battery pack
  • Environmentally Preferable:
  • Pk:
    1