Surgical masks, Foliodress® mask comfort special
HART9921421
- Type:With ties
- Colour:Green
- Pk:50
Comfortable extra wide surgical face masks made from special combination of soft and hypoallergenic non-woven materials.
- Integrated filter for a high bacterial filtration efficiency and low breathing resistance
- Long integrated softly cushioned nosepiece
- Stitched ties due to the extra wide cut
- Free from natural rubber latex, fragrance free and durable
Suitable for use in all surgical areas, like isolation wards, intensive care departments, delivery rooms, premature and newborn infant wards.
: EN 14683 Type II