ContactUsLinkComponent
Strips of 12 PCR tubes and cap strips
Strips of 12 PCR tubes and cap strips
Catalog # DELT4095.4N
Supplier:  DELTALAB
Strips of 12 PCR tubes and cap strips
Catalog # DELT4095.4N
Supplier:  DELTALAB
Specifications
  • Packed:
    80 strips/case
  • Capacity:
    0,2 ml
  • Colour:
    Natural
  • Description:
    Strips of 12 tubes and 12 caps
  • Pk:
    80
Specifications
About this item
Tubes made of PP.
  • RNAse, DNAse and pyrogen free
  • Excellent chemical resistance
  • Excellent chemical resistance
Cap and tube strips allow easy handling and inventory of tubes and caps. It also prevent cross-contamination from tube to tube.