Order Entry
Czech Republic
ContactUsLinkComponent
Racks for cryovials and microtubes with lid
Racks for cryovials and microtubes with lid
Catalog # 212-8450
Supplier:  DELTALAB
undefined
Racks for cryovials and microtubes with lid
Catalog # 212-8450
Supplier:  DELTALAB

Specifications

  • No. of Holes:
    50
  • Description:
    Styrofoam racks for 1,5/2,0 ml microtubes or 1,2 ml cryovials, with lid
  • Seq:
    01
  • W×D×H:
    210×107×65 mm
  • Environmentally Preferable:
  • Pk:
    10

Specifications

About this item

Styrofoam, white

  • Designed for long-term storage of microtubes and cryovials in the freezer
  • Alpha-numerically matrixed for better sample location
  • Stackable and lightweight