About this item
Extremely conformable, vinyl cleanroom tape with synthetic rubber, clean-peel adhesive; ideal for colour coding, sealing and marking on curved and irregular surfaces in cleanroom maintenance and support areas.
- Clean peel from wafer cartridges and cleanroom work surfaces
- Synthetic rubber adhesive ensures a clean peel from most cleanroom surfaces such as polyethylene, plastics and stainless steel
- Matte finish can be written on with a cleanroom pen
- Ideal for taping and sealing curved and irregular surfaces
- Special processing and packaging techniques ensure low particles and extractable ion counts for applications in and around the cleanroom
- Wound on a 76 mm (3") plastic core
Length: 33 m
Available in a variety of colours and widths.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- MICRCR100PC-1DB
- 811-0205
- MICRCR100-34TRIRM
- MICRCR100PC-1RE
- MICRCR100PC-14WH
- MICRCR100PC-2DB
- MICRCR100PC-1LG
- MICRCR100PC-12RE
- MICRCR100PC-12WHPK
- MICRCR100PC-2LB
- MICRCR100PC-3WH
- MICRCR100PC-4WH
- MICRCR100PC-3RE
- MICRCR100PC-3DG
- MICRCR100PC-12TR
- MICRCR100PC-12DG
- MICRCR100PC-15DB
- MICRCR100PC-2TR
- MICRCR100PC-14RE
- 819-0020
- MICRCR100PC-1TR
- MICRCR100PC-2DG
- MICRCR100PC-38DB
- MICRCR100PC-3DB
- MICRCR100PC-2LG
- MICRCR100PC-15LB
- MICRCR100PC-38YE
- 811-0206
- MICRCR100PC-1LB
- MICRCR100PC-1DG
- MICRCR100PC-2WH
- MICRCR100PC-1OR
- MICRCR100PC-1YE
- MICRCR100PC-2RE
- MICRCR100PC-2YE
- MICRCR100PC-14YE
- MICRCR100-12WHIRM
- MICRCR100PC-34WH
- MICRCR100PC-38TR
- MICRCR100PC-38WH
- MICRCR100PC-12WH
- MICRCR100PC-38DG
- MICRCR100PC-4TR
- MICRCR100PC-15YE
- MICRCR100PC-15WH
- MICRCR100PC-15TR
- Environmentally Preferable:
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient