Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Boxes
Transport Boxes
Storage and Transportation Box
Storage and Transportation Box
Storage and Transportation Box
Catalog # DALL1002864
Supplier:
HONEYWELL SAFETY
Expand image
Storage and Transportation Box
Catalog # DALL1002864
Supplier:
HONEYWELL SAFETY
Specifications
Description:
Storage box for fall protection equipment, PVC, 1,7 kg
W×D×H:
500×300×200 mm
Environmentally Preferable:
Pk:
1
Specifications
About this item
Product Details
Product Details