Order Entry
Czech Republic
ContactUsLinkComponent
 
Cell culture media, Ham's F14 (F-14 Nutrient Medium)
Catalog # L0138-500
Supplier:  Biowest
CAS Number:  
Cell culture media, Ham's F14 (F-14 Nutrient Medium)
Catalog # L0138-500
Supplier:  Biowest
CAS Number:  

Specifications

  • Media Format:
    Liquid
  • Pk:
    500 ml
  • Description:
    Ham’s F14, with 6 g/L glucose and 1 mg/L ATP
  • Seq:
    75

Specifications

About this item

Ham's F14 medium was developed from Ham's F12 medium, which was originally developed for the serum-free clonal growth of Chinese Hamster Ovary (CHO) cells, lung cells and mouse L-cells. Ham's F14 contains a double concentration of amino acids compared to the Ham's F12 medium. This product is also supplemented with calcium chloride and ascorbic acid.