Urine and biological specimen containers, with ribbed cap
Supplier: DELTALAB
Certificates
About this item
HDPE container and white ribbed cap.
- Convenient handle for an easy single-handed use
- Features a pharmaceutical grade joint, which eliminates the need for an insert plug, allows a better handling and provides a water tight seal
- Containers are graduated in 50 ml increments up to 2 L
Ideal for 24 hour urine collection and any other liquid samples.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together