Examination gloves, latex, Unigloves® Comfort
Supplier: UNIGLOVES AERTZE UND KLINIKBED
Certificates
About this item
These disposable gloves are suitable for use in non hazardous environments. Ideal for use in medical examinations.
- Non sterile, powder-free
- Micro-roughened palm for improved grip
- Rolled cuff
- Double chlorinated
Certifications: EN 455 (1-4), EN ISO 374-1 Type C, EN ISO 374; AQL 1,5
Specifications
- Catalog No:
- 112-1703
- 112-1714
- 112-1704
- 112-1702
- 112-1585
- Model:
- Comfort
- Comfort
- Comfort
- Comfort
- Comfort
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Micro-textured
- Micro-textured
- Micro-textured
- Micro-textured
- Micro-textured
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Cuff thickness:
- 0,12 - 0,15 mm
- 0,12 - 0,15 mm
- 0,12 - 0,15 mm
- 0,12 - 0,15 mm
- 0,12 - 0,15 mm
- Finger thickness:
- 0,18 - 0,22 mm
- 0,18 - 0,22 mm
- 0,18 - 0,22 mm
- 0,18 - 0,22 mm
- 0,18 - 0,22 mm
- Palm thickness:
- 0,15 - 0,18 mm
- 0,15 - 0,18 mm
- 0,15 - 0,18 mm
- 0,15 - 0,18 mm
- 0,15 - 0,18 mm
- EN norm:
- EN 455 (1-4), EN ISO 374-1 Type C, EN ISO 374
- EN 455 (1-4), EN ISO 374-1 Type C, EN ISO 374
- EN 455 (1-4), EN ISO 374-1 Type C, EN ISO 374
- EN 455 (1-4), EN ISO 374-1 Type C, EN ISO 374
- EN 455 (1-4), EN ISO 374-1 Type C, EN ISO 374
Specifications
Frequently Bought Together