Mini eye showers for tap attachment
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Certificates
About this item
These mini eye showers are made from chrome plated brass.
Model 2000: Water flows to the perlator for washing hands, pressing the knob (red) then transfers the water flows through the eye nozzles.
Model 2001: Water flows through the eye nozzles, pressing the knob (red) then transfers the water flow to the Perlator for washing hands.
Delivery: Supplied with M22 adapters.
Frequently Bought Together