High vacuum angle valves, with sealed bellows, VA series
Supplier: Welch by Gardner Denver
Certificates
About this item
These high vacuum angle valves feature a very long life bellows-seal made from stainless steel.
- High conductance
- Leak rate 10-9 mbar L/s
Frequently Bought Together