ContactUsLinkComponent
Special cleaning wipes
Special cleaning wipes
Catalog # KOEHK-1560
Special cleaning wipes
Catalog # KOEHK-1560
Specifications
  • Colour:
    White
  • L×W:
    400×380 mm
  • Pk:
    40
Specifications
About this item
Non woven wipes for universal use.
  • Lint-free
  • Reusable
  • Ideal for cleaning goggles
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904