ContactUsLinkComponent
Pipette stand
Pipette stand
Catalog # KOEHA-1650
undefined
Pipette stand
Catalog # KOEHA-1650
Specifications
  • Colour:
    Light grey/black
  • Description:
    Pipette stand with two rows
  • W×D×H:
    280×100×210 mm
  • Pk:
    1
Specifications
About this item
Universal pipette stand made of rigid PVC. Accommodates up to 9 µl pipettes from various manufacturers.
  • Stable
  • Ergonomically tilted