Cryo-Clear™ laser cryo labels
Supplier: DIVERSIFIED BIOTECH
Certificates
About this item
These Cryo-Clear™ vinyl cryo labels are specially top-coated to accept laser printing and other markings.
- Temperature resistant from −196 to 70 °C
- Chemically inert, resist most organic solvents and caustic agents
- Adhere to plastics, glass and metals without cracking, peeling or degrading
Cryo-Clear™ labels are made with a flexible, vinyl label material and permanent acrylic adhesive. This unique combination expands and contracts together as temperatures fluctuate resulting in a strong and sustained adhesive bond. They remain fully adhered down to −196°C (liquid and vapor phase nitrogen).
Frequently Bought Together