ContactUsLinkComponent
Weighing boats
Weighing boats
Catalog # DELT1911103
Supplier:  DELTALAB
Weighing boats
Catalog # DELT1911103
Supplier:  DELTALAB
Specifications
  • W×D×H:
    41×41×8 mm
  • Pk:
    500
Specifications
About this item
PS, square, white.
  • Resistant to weak acids, water based solutions and alcohol
  • Can be folded for easy pouring
  • Suitable for microwave