O tomto produktu
Several nut lengths and head styles to fit into a variety of applications, designed to connect ¹/₁₆" OD tubing to 10-32 coned ports, hold up to 9000 psi (620 bar).
The dual compression created by the specially designed nut and ferrule enables the SealTight fittings system to outperform standard finger tightened fittings. The forward cone of
the SealTight Ferrule provides gripping power and a leak-free seal via conventional compression by the receiving port. The slotted end creates the second compression zone in conjunction with a SealTight nut, to be used with the F-192 ferrule.
A wide variety of fitting head styles are available for various space constraints. This fittings system is also interchangeable with the two-piece RheFlex® fittings system.