ESD occupational shoes, clogs, Nature, 4040/4045/4050/4055
Supplier: Abeba
Certificates
About this item
ESD shoe made from smooth or perforated leather with non foldable, adjustable heel strap and a rubber sole (resistant to fuels, oils, acids and bases).
- Skin friendly leather coated, cork integrated insole with toe gripper
- Sanitized® treatment reduces odour and bacterial growth
- Slip resistant special rubber sole with shock-absorbent heel
- Adjustable instep area with buckle fastening
Colour: White, black or dark blue
Colour: White, black or dark blue
Certifications: EN 61340 (ESD); TÜV GS DIN 4843/100 § 3.1v.8/93, EN ISO 20347:2012; OB, FO, A, SRC
Frequently Bought Together