ESD occupational shoes, clogs, Rubber, 5200/5250
Dodavatel: Abeba
Certifikáty
O tomto produktu
ESD clogs made from smooth leather, a foldable, adjustable heel strap, and a rubber sole with cross/circle profile, resistant to acids, bases (cleaning agents) and fuels.
- Sanitized® treatment reduces odour and bacterial growth
- Non slip special rubber sole
Colour: White or blue
Certifications: EN 61340 (ESD); TÜV GS DIN 4843/100 § 3.1v.8/93, EN ISO 20347:2012; OB, A, FO, WRU, SRC.
Frequently Bought Together