
4-Bromo-3-chlorobenzotrifluoride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:402-04-0
:
Formula:C₇H₃BrClF₃
MDL Number:MFCD00673985
:259.45 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together