
3-Fluoro-2-nitrophenol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:385-01-3
:
Formula:C₆H₄FNO₃
MDL Number:MFCD07368753
:157.10 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together