Order Entry
Czech Republic
ContactUsLinkComponent
L(+)-Ornithine hydrochloride ≥99%
L(+)-Ornithine hydrochloride ≥99%
Catalog # 0958-500G
Supplier:  VWR Chemicals
CAS Number:  
undefined
L(+)-Ornithine hydrochloride ≥99%
Catalog # 0958-500G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Ammonium:
    = 0.1%
  • Heavy Metals:
    = 10 ppm
  • Iron:
    = 10 ppm
  • Loss on Drying:
    = 0.5%
  • Purity:
    = 99%
  • Specific Rotation (Dried, c=4, 6N HCl):
    23 °- 25 °
  • Sulphate:
    = 0.02%

Specification Test Results

About this item

 :
 :168.62 g/mol
EINECS:221-678-6
CAS Number:3184-13-2
Formula:C₅H₁₂N₂O₂·HCl
MDL Number:MFCD00064562