
4-Fluorobenzamidine hydrochloride 95%
Catalog # H61570.MD
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
Specifications
About this item
:
:174.61 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:456-14-4
Formula:C₇H₈ClFN₂
MDL Number:MFCD04114421