Hydrochloric acid 35% for synthesis
Catalog # SCARAC07371000
Supplier: Scharlau
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.18 g/cm³ (20 °C)
ADR:8,II
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324