
1-Ethyl-2,3-dimethylimidazolium chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:92507-97-6
:
Formula:C₇H₁₃ClN₂
MDL Number:MFCD03093290
:160.65 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together