Sodium chloride 8.7 g/l in aqueous solution
Catalog # REGENACL087WV5
Supplier: REAGECON
CAS Number:
Specifications
- Pk:4 L
- Seq:5162
Specifications
About this item
:
:58.44 g/mol
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477