Order Entry
Czech Republic
ContactUsLinkComponent
Tetrafluoroterephthalonitrile ≥98%
Toxic
Irritant
Tetrafluoroterephthalonitrile ≥98%
Catalog # H61326.14
CAS Number:  
Toxic
Irritant
Tetrafluoroterephthalonitrile ≥98%
Catalog # H61326.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Melting Pt:197…199 °C
EINECS:217-397-3
ADR:6.1,III
 :200.1 g/mol
UN:3439
 :GHS06
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1835-49-0
Formula:C₈F₄N₂
MDL Number:MFCD00001776