Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
Health
Iodine concentrate for dilution to 0.25 M (0.5 N), Fluka™
Catalog # 38084-1EA
CAS Number:  
Irritant
Health
Iodine concentrate for dilution to 0.25 M (0.5 N), Fluka™
Catalog # 38084-1EA
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
 :253.81 g/mol
EINECS:231-442-4
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:7553-56-2
Formula:I₂
MDL Number:MFCD00011355