1,2-Diphenoxyethane, Sigma-Aldrich®
Catalog # SIAL140287-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:214.26 g/mol
:M-Clarity™ quality level = MQ100
EINECS:203-224-9
CAS Number:104-66-5
Formula:C₁₄H₁₄O₂
MDL Number:MFCD00003039