

Cerium(IV) sulphate hydrate ≥99% (REO, rare earth oxide basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:95838-16-7
:
Formula:CeO₈S₂·H₂O
MDL Number:MFCD00149427
:332.24 g/mol (anhy)
Melting Pt:195 °C (dec.)
Density:3.01
Storage Temperature:Ambient
EINECS:237-029-5
ADR:5.1,III
:GHS05
:GHS03
Related Information
Frequently Bought Together