Order Entry
Czech Republic
ContactUsLinkComponent
Potassium stannate trihydrate
Potassium stannate trihydrate
Catalog # ABCR106619
Supplier:  abcr
CAS Number:  
Potassium stannate trihydrate
Catalog # ABCR106619
Supplier:  abcr
CAS Number:  

Specifications

  • Pk:
    1 KIT

Specifications

About this item

 :
 :298.94 g/mol (244.89 anhy)
EINECS:235-255-9
CAS Number:12125-03-0
Formula:K₂O₃Sn·3H₂O
MDL Number:MFCD00150391