Order Entry
Czech Republic
ContactUsLinkComponent
3-(2,4-Dichlorophenoxy)propionic acid
3-(2,4-Dichlorophenoxy)propionic acid
Catalog # EHERC12510000
Supplier:  EHRENSTORFER
CAS Number:  
3-(2,4-Dichlorophenoxy)propionic acid
Catalog # EHERC12510000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :235.07 g/mol
EINECS:204-390-5
CAS Number:120-36-5
Formula:C₉H₈Cl₂O₃
MDL Number:MFCD00002645