1-Acetoxy-1,3-butadiene (mixture of cis and trans isomeres), Sigma-Aldrich®
Catalog # SIAL220868-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
EINECS:216-159-6
CAS Number:1515-76-0
Formula:C₆H₈O₂
MDL Number:MFCD00008714