4-(Difluoromethoxy)iodobenzene
Catalog # ABCRF02847R.0025
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:2,5 g
Specifications
About this item
:
:270.0153 g/mol
CAS Number:128140-82-9
Formula:C₇H₅F₂IO
MDL Number:MFCD00085008