Order Entry
Czech Republic
ContactUsLinkComponent
2,2-Dimethylbutyryl chloride ≥98%
Corrosive
Flammable
2,2-Dimethylbutyryl chloride ≥98%
Catalog # ABCRAB147407.0250
Supplier:  abcr
CAS Number:  
Corrosive
Flammable
2,2-Dimethylbutyryl chloride ≥98%
Catalog # ABCRAB147407.0250
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

About this item

 :
 :134.61 g/mol
Boiling Pt:133 °C
Density:0.980
Flash Pt:29 °C (84 °F)
EINECS:227-478-5
ADR:8,II
 :GHS05
 :GHS02
Storage Temperature:Ambient
CAS Number:5856-77-9
Formula:C₆H₁₁ClO
MDL Number:MFCD03093071