L(+)-Isoleucinol, Sigma-Aldrich®
Catalog # SIAL190527-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:117.19 g/mol
:M-Clarity™ quality level = MQ100
EINECS:246-371-4
CAS Number:24629-25-2
Formula:C₆H₁₅NO
MDL Number:MFCD00004731