
L(+)-Isoleucinol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:24629-25-2
:
Formula:C₆H₁₅NO
MDL Number:MFCD00004731
:117.19 g/mol
Boiling Pt:97 °C (14 mmHg)
Melting Pt:36…39 °C
Flash Pt:100 °C
Storage Temperature:Ambient
EINECS:246-371-4
:GHS07
Specification Test Results
- Appearance:Colorless to white to light yellow Low melting mass and/or chunks or liquid
- Infrared spectrum:Conforms
- Melting point:35 to 40 °C
- Specific optical rotation:+3.5° to +5.5° (20 °C, 589 nm) (c=1.6, C2H5OH)
- Titration with HClO4:≥96.0 %
Specification Test Results
Frequently Bought Together