(R)-(+)-2-Hydroxy-3-phenylpropionic acid, Sigma-Aldrich®
Katalogové číslo SIAL376906-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:166.1739 g/mol
:M-Clarity™ quality level = MQ100
EINECS:230-803-3
CAS Number:7326-19-4
Formula:C₉H₁₀O₃
MDL Number:MFCD00078062