(R)-(+)-2-Hydroxy-3-phenylpropionic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7326-19-4
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00078062
:166.1739 g/mol
EINECS:230-803-3
Related Information
Frequently Bought Together