
4,4'-Difluorobenzhydryl chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:27064-94-4
:
Formula:C₁₃H₉ClF₂
MDL Number:MFCD00044329
:238.66 g/mol
Boiling Pt:126…128 °C (1 mmHg)
Density:1.280
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:248-201-4
ADR:8,III
:GHS05
Related Information
Frequently Bought Together