Order Entry
Czech Republic
ContactUsLinkComponent
 
1-(4-Nitrophenylazo)-2-naphthol, Supelco®
Catalog # 40446-100MG
Supplier:  Merck
CAS Number:  
undefined
1-(4-Nitrophenylazo)-2-naphthol, Supelco®
Catalog # 40446-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :293.28 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:229-093-8
CAS Number:6410-10-2
Formula:C₁₆H₁₁N₃O₃
MDL Number:MFCD00003908