Order Entry
Czech Republic
ContactUsLinkComponent
Methyl-4-(hydroxymethyl)benzoate 98%
 
Methyl-4-(hydroxymethyl)benzoate 98%
  ACRO330840100
 :  
undefined
 
Methyl-4-(hydroxymethyl)benzoate 98%
  ACRO330840100
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

 

 

  • Appearance:
    White Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    45 to 50 °C
  • GC:
    ≥97.5 %

 

 

 :
 :45…50 °C
 :>110 °C
 :166.18 g/mol
 :230-017-0
 :GHS07
 :Ambient
 :6908-41-4
 :C₉H₁₀O₃
 :MFCD00009984