N-Boc-L-phenylalaninol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:66605-57-0
:
Formula:C₁₄H₂₁NO₃
MDL Number:MFCD00076976
:251.3 g/mol
Melting Pt:95…98 °C
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together