
2-Nitrobenzenesulphenyl chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7669-54-7
:
Formula:C₆H₄ClNO₂S
MDL Number:MFCD00007128
Melting Pt:72…75 °C
EINECS:231-644-2
ADR:8,II
:189.62 g/mol
:GHS05
Related Information
Frequently Bought Together