Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Bis(4-aminophenoxy)benzene
1,3-Bis(4-aminophenoxy)benzene, Sigma-Aldrich®
1,3-Bis(4-aminophenoxy)benzene, Sigma-Aldrich®
1,3-Bis(4-aminophenoxy)benzene, Sigma-Aldrich®
SIAL476323-100G
:
Merck
:
1,3-Bis(4-aminophenoxy)benzene, Sigma-Aldrich®
SIAL476323-100G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
100 g
Seq:
3401
:
:
292.3300 g/mol
:
M-Clarity™ quality level = MQ100
:
2479-46-1
:
C₁₈H₁₆N₂O₂
:
MFCD00039154