

2-(Methylthio)ethylamine 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18542-42-2
:
Formula:C₃H₉NS
MDL Number:MFCD00014825
:91.18 g/mol
Boiling Pt:146…148 °C
Density:0.974
Flash Pt:36 °C (96 °F)
Storage Temperature:Ambient
EINECS:242-412-5
ADR:8,II
:GHS05
:GHS02
Related Information
Frequently Bought Together