

1,6-Diaminohexane 60% (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:124-09-4
:
Formula:C₆H₁₆N₂
MDL Number:MFCD00008243
:116.21 g/mol
Flash Pt:116 °C
Storage Temperature:Ambient
EINECS:204-679-6
UN:1783
ADR:8,II
:GHS07
:GHS05
Specification Test Results
- Appearance:Clear solution
- Color scale:≤20 APHA
- Strength:59 to 61 wt%
Specification Test Results
Frequently Bought Together