Order Entry
Czech Republic
ContactUsLinkComponent
4,6-Dichloro-2-(methylthio)pyrimidine 98%
Corrosive
4,6-Dichloro-2-(methylthio)pyrimidine 98%
Catalog # ACRO163030050
CAS Number:  
undefined
Corrosive
4,6-Dichloro-2-(methylthio)pyrimidine 98%
Catalog # ACRO163030050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to yellow to orange Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    38 to 43 °C
  • GC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :195.07 g/mol
Boiling Pt:135…136 °C (14 mmHg)
Melting Pt:38…42 °C
Flash Pt:>112 °C
Storage Temperature:Ambient
EINECS:228-577-6
UN:3263
ADR:8,II
 :GHS05
CAS Number:6299-25-8
Formula:C₅H₄Cl₂N₂S
MDL Number:MFCD00006086