1-Acetylpyrene ≥97%
Catalog # B22483.88
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:2,5 g
Specifications
About this item
:
:244.29 g/mol
Melting Pt:86…89 °C
Storage Temperature:Ambient
CAS Number:3264-21-9
Formula:C₁₈H₁₂O
MDL Number:MFCD00021663