Order Entry
Czech Republic
ContactUsLinkComponent
1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazole-5-boronic acid pinacol ester 95%
1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazole-5-boronic acid pinacol ester 95%
Catalog # H27043.03
CAS Number:  
1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazole-5-boronic acid pinacol ester 95%
Catalog # H27043.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :324.31 g/mol
Storage Temperature:Ambient
CAS Number:903550-12-9
Formula:C₁₅H₂₉BN₂O₃Si
MDL Number:MFCD09037502